Skip to Content
Merck
All Photos(3)

Documents

EP4987000410

Eppendorf® Repeater E3/E3x

E3x single-channel, with charging cable, charger stand, and Combitips advanced assortment pack (1 Combitip of each size), volume 1—50 mL, AC/DC input 100—240 V, 50—60 Hz, US 2-pin plug

Synonym(s):

Adjustable volume pipette

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.15

description

incl Combitips advanced assortment pack (1 Combitip of each size)

feature

adjustable
electronic
no. of channels Single Channel

packaging

pack of 1

manufacturer/tradename

Eppendorf® 4987000410

volume

1—50 mL

battery power

lithium (incl Battery, charging cable and charger stand)

Looking for similar products? Visit Product Comparison Guide

General description

Repeater E3x bundle incl. charger stand, 1-channel, with charging cable and Combitips"] advanced assortment pack (1 Combitips® advanced tip of each size), 1 µL – 50 mL
The Multipette E3/E3x in combination with the Combitips® advanced creates a versatile dispensing system that functions according to the positive displacement principle and is therefore capable of pipetting any liquid while maintaining the highest standards for accurate results. The motor driven dispenser gives you full volume control, improved accuracy and reproducibility while reducing physical strain during use. The repetitive dispensing modes contribute to reducing fatigue by decreasing the number of movements for the user; filling the tip once allows dispensing up to 100 times. The system is an essential lab tool ideal for all liquid handling applications, accommodating more than 5,000 different dispensing volumes between 1 μL to 50 mL.

Features and Benefits

  • Electronic positive displacement dispenser with several functions e.g. for dilution series, titration, automatic dispensing into microplates and vessel series
  • Automatic Combitips® advanced pipette tip recognition: Eliminates time-consuming volume calculations, avoiding incorrect dispensing volumes
  • Positive displacement principle: Precise, accurate and contamination-free dispensing of viscous solutions (e.g. glycerol), liquids with high vapor pressure (e.g. ethanol) or toxic, radioactive and other hazardous samples
  • Motor-driven operation: Reduces the human error to a minimum and minimizes risks of repetitive strain injuries (RSI)
  • Speed adjustment: Program aspiration and dispensing speed to meet your throughput, precision and accuracy goals while also preventing splashing
  • Ergonomic one-button tip ejector: For one-handed operation and contact-free Combitips® advanced pipette tip ejection
  • Increase your efficiency when filling plates and series of tubes: Fill the tip once and dispense up to 100 times without a refill
  • Versatile system for different applications: 1 µL to 50 mL dispensing range providing up to 5,000 dispensing volumes with increments as low as 100 nanoliters
  • 9 display languages: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish
  • Eco-conscious production: Multipette E3/E3x and Combitips® advanced tips are ACT®-labeled to help you make an informed choice when it comes to sustainability

Legal Information

ACT is a registered trademark of Merck KGaA, Darmstadt, Germany
Combitips is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Repeater is a trademark of Brinkmann Instruments, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service