Merck
All Photos(4)
Key Documents
EP4926000018
Eppendorf® Reference® 2 Variable Volume Pipettor
8-channel, incl. epT.I.P.S.® Box, volume 0.5-10 μL, gray
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
material
gray
feature
8-channel
mechanical
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 4922000013 (Global)
volume
0.5-10 μL
Related Categories
General description
The Reference 2 is the successor to the highly successful Reference pipette, and enhances the range′s already high standards with a new design, reduced weight and operating forces and multi-channel versions. As the premium product in Eppendorf′s pipette portfolio, the Reference 2 provides the most accurate results possible alongside robust, reliable handling and optimum user safety.
These levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducability, Eppendorf′s new Reference 2 is an ideal instrument for these liquid handling applications.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)