Key Documents
EP4720000011
Eppendorf® Varipette 4720
with coninuous volume adjustment 1-10 mL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
with coninuous volume adjustment 1-10 mL
manufacturer/tradename
4720000011
Eppendorf®
General description
Handle challenging liquid handling tasks with ease! The Varipette 4720 is a large volume, positive displacement pipette that is optimized for lab work with viscous or volatile liquids. Ideal for safely aspirating liquids from large bottles or tall, narrow vessels. Super easy, one-handed operation significantly speeds up pipetting tasks – simply press down the Varipettor operating lever to aspirate liquid from the reservoir; press the lever again and the liquid will be dispensed into your sample vessel.
Features and Benefits
  • Mechanical positive displacement pipette designed for precise pipetting of liquids with high vapor pressure or viscosity
  • Volume range: 1 mL to 10 mL, adjustable in 10 µL increments for accurate volume setting via 4-digit volume display
  • Fast pipetting and convenient operation due to unique one-hand operation.
  • Liquid automatically aspirates and dispenses by simply pressing the operating lever
  • Eppendorf Varitips P is a syringe-style, positive displacement tip that allows for accurate pipetting of viscous solutions or liquids with high-vapor pressure
  • The Eppendorf Varitips S system, consisting of the dispensing part and Varitips, is optimized for aspirating liquids from large bottles or tall, narrow vessels
  • A valve for use with the Varitips S system ensures drip-free dispensing of volatile solutions
  • The graduation on Varitips ensures accurate dispensing of non-aqueous liquids
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)