Key Documents
EP3880001174
Eppendorf® IsoPack and IsoRack set
for 1.5-2.0 mL tubes, -21degC
About This Item
UNSPSC Code:
41102404
NACRES:
NC.21
Pricing and availability is not currently available.
manufacturer/tradename
Eppendorf® 3880001174
Features and Benefits
  • Eppendorf® IsoPack and IsoRack set, includes one IsoPack and one IsoRack.
  • Ideal for cooling, transporting, and storing deep-frozen samples.
  • Prevents enzymes from becoming inactive as well as for PCR sample preparation, sample transport and slow thawing of frozen samples.
  • Dry incubation technology reduces contamination risk associated with classical icebath incubation for safe sample handling.
  • Holds up to 24 μ tubes for broad flexibility.
  • One version of IsoPack available: Blue maintains -21 °C for up to 3 hour.
  • Racks are stackable, autoclavable and can be centrifuged in the MTP rotor.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)