Skip to Content
Merck
All Photos(1)

Documents

EP022364111

Eppendorf® Flex-Tubes Microcentrifuge Tubes

clear, capacity 1.5 mL, pack of 500 ea

Synonym(s):

eppendorfs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703

material

clear
polypropylene
polypropylene cap

description

push fit

sterility

non-sterile

packaging

pack of 500 ea

manufacturer/tradename

Eppendorf® 022364111

parameter

30,000 × g max. RCF

D

10.7 mm

capacity

1.5 mL

Related Categories

General description

Flex-Tube®, 1.5 mL, Eppendorf Quality, colorless, 500 pcs.
  • Flex-Tube lid is easy to open and close for ergonomic operation
  • Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes
  • g-Safe®: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
  • Graduation for visual control of the pipetted volume
  • Frosted lid and surface on the side for easy labeling
  • Ensured functionality from -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)
  • Available in Eppendorf Quality and lot-certified PCR clean purity grade

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Flex-Tube is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service