Skip to Content
Merck
All Photos(1)

Key Documents

EP0030605802

Eppendorf® Microplate 96/U-PP, 96 wells

white border (wells black), PCR clean, polypropylene, round bottom black wells, white polypropylene, pkg of 240 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15
material:
round bottom black wells
white polypropylene
size:
96 wells
Pricing and availability is not currently available.

material

round bottom black wells
white polypropylene

packaging

pkg of 240 ea

manufacturer/tradename

Eppendorf® 0030605802

technique(s)

PCR: suitable

size

96 wells

Looking for similar products? Visit Product Comparison Guide

General description

Pipette, check and recover your samples easily with Eppendorf round bottom Microplates. The easy-to-read OptiTrack alphanumeric matrix label for identifying each well speeds up filling and significantly reduces the risk of pipetting errors.

Application

  • sample storage and preparationassays that require high resistance against temperature or solvents
  • active ingredient screening
  • combinatorial chemistry
  • storage of cDNA or genomic banks
  • protein analysis
  • genotyping
  • storage of siRNA or oligonucleotide libraries
  • preparation of tissue and plant lysates

Features and Benefits

  • [TM=“Eppendorf”] Microplates consist of polypropylene wells which enhance sample visibility. In addition, polypropylene also ensures resistance towards chemicals, mechanical stressors and high temperatures.
  • A raised well rim and a smooth surface facilitates secure sealing.
  • [TM=“Eppendorf”] Microplates can withstand centrifugal forces up to six thousand times the gravitational force.
  • The [TM=“OptiTrack”] matrix feature enables faster well identification and reduces pipetting error.
  • An optimized [TM=“RecoverMax”] well geometry minimizes the remaining volume and ensures smooth mixing properties.
  • [TM=“Eppendorf”] Microplates function efficiently over a wide temperature range, ranging from -86 °C to 100 °C.
  • These microplates can be autoclaved at 121 °C for 20 min.

  • Eppendorf Microplates consist of polypropylene wells which enhance sample visibility. In addition, polypropylene also ensures resistance towards chemicals, mechanical stressors and high temperatures.
  • A raised well rim and a smooth surface facilitates secure sealing.
  • Eppendorf Microplates can withstand centrifugal forces up to six thousand times the gravitational force.
  • The OptiTrack matrix feature enables faster well identification and reduces pipetting error.
  • An optimized RecoverMax well geometry minimizes the remaining volume and ensures smooth mixing properties.
  • Eppendorf Microplates function efficiently over a wide temperature range, ranging from -86 °C to 100 °C.
  • These microplates can be autoclaved at 121 °C for 20 min.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service