Key Documents
EP0030601670
Eppendorf® Microplate 96/V-PP, 96 wells
gray polypropylene, conical bottom white wells, PCR clean, pkg of 80 ea
About This Item
UNSPSC Code:
41122107
NACRES:
NB.15
material:
conical bottom white wells
gray polypropylene
size:
96 wells
sterility:
sterile; autoclaved
Pricing and availability is not currently available.
material
conical bottom white wells
gray polypropylene
sterility
sterile; autoclaved
packaging
pkg of 80 ea
manufacturer/tradename
Eppendorf® 0030601670
technique(s)
PCR: suitable
size
96 wells
General description
Pipette, control and recover your samples with ease using Eppendorf Microplates! Designed with the easy-to-read alphanumeric OptiTrack matrix label to identify individual wells, speed up the filling process and significantly reduce the risk of pipetting errors. Eppendorf Microplates are made with high-transparency polypropylene ensuring quick and easy location of samples
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)