Skip to Content
Merck
All Photos(1)

Key Documents

EP0030125185

Sigma-Aldrich

Eppendorf® Microtube 3810X

capacity 1.5 mL, green, pkg of 1000 ea

Synonym(s):

Eppendorfs, Eppendorf tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.02
Pricing and availability is not currently available.

material

cap (push fit)
green
polypropylene

sterility

non-sterile

packaging

pkg of 1000 ea

manufacturer/tradename

Eppendorf® 0030125185

capacity

1.5 mL

maximum volume

1.5 mL

suitability

suitable for (g-safe® centrifugation stability)

Looking for similar products? Visit Product Comparison Guide

General description

The easy-to-open and close design of the Eppendorf® 3810X tubes offers convenient, ergonomic and reliable one-hand operation. Whether sample preparation, centrifugation or storage, the Eppendorf® 3810X tubes fit perfectly into your workflows.

Features and Benefits

  • Tube lid is easy to open and close for ergonomic operation
  • Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes g-Safe®: Exceptional centrifugation stability up to 30,000 x g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
  • Graduation for visual control of the pipetted volume
  • Frosted lid and surface on the side for easy labeling
  • Ensured functionality from -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service