Skip to Content
Merck
All Photos(1)

Documents

EP0030122321

Eppendorf® Tubes 5.0 mL

sterile, 5.0 mL, colorless, pkg of 200 tubes (2 bags of 100 tubes), with screw cap

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.22

material

conical bottom polypropylene

sterility

sterile

feature

with screw cap

packaging

pkg of 200 tubes (2 bags of 100 tubes)

manufacturer/tradename

Eppendorf® 0030122321

measuring range

5.0 mL

parameter

25000 × g max. RCF

technique(s)

PCR: suitable

D

17 mm

color

colorless

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® 5.0 mL Tubes are made of high-quality, transparent polypropylene (PP).
  • Tubes are pyrogen-, DNase-, RNase-, human and bacterial DNA-free for accurate test results.
  • Includes a flattened screw cap with grooved sides for optimal grip.
  • Conical shape for use in many adapters and racks.
  • Screwcap tubes can withstand temperatures between -86 °C to 100 °C and centrifugation speed up to 25,000 x g.
Eppendorf 5.0 mL tube′s conical shape is designed to be compatible with most accessories for common conical 15 mL tubes. This allows you to continue using many of your existing adapters and racks!Eppendorf′s "missing link": the Eppendorf Tube 5.0 mL, fills the gap between existing tube versions and enables the simple and safe processing of sample volumes up to 5.0 mL.

Application

  • Preparation of mastermixes and buffers
  • Cell and tissue lysis
  • Convenient and safe cell culture applications.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service