Skip to Content
Merck
All Photos(1)

Key Documents

EMU058651

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Aof2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACTGCCCTCTGCAAGGAATATGATGAATTAGCTGAAACACAAGGAAAGCTAGAAGAAAAACTTCAAGAATTGGAAGCCAATCCCCCAAGTGATGTATACCTCTCATCAAGAGACAGACAAATACTTGACTGGCATTTTGCAAATCTTGAATTTGCCAACGCCACACCTCTCTCTACCCTCTCTCTTAAACATTGGGATCAGGATGATGACTTTGAGTTTACTGGAAGCCACCTGACAGTAAGGAATGGCTACTCATGTGTGCCTGTGGCTTTAGCTGAAGGCTTGGACATTAAACTGAACACAGCAGTGCGGCAGGTTCGCTACACAGCCTCAGGATGTGAAGTGATTGCTGTGAACACACGTTCCACAAGTCAAACCTTTATTTATAAGTGTGATGCAGTTCTCTGTACACTTCCTTTGGGAGTGTTGAAGCAGCAGCCACCAGCTGTTCAGTTTGTGCCACCTCTTCCTGAGTGGAAAACATCTGCAGTCCAAAGGATGGGATTTGGCAACCTTAACAAGGTGGTGTTATGCTTTGACCGTGTGTTCTGGGACCCAAGT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa M Singh et al.
Neuro-oncology, 17(11), 1463-1473 (2015-03-22)
Glioblastoma (GBM) is the most common and aggressive form of brain cancer. Our previous studies demonstrated that combined inhibition of HDAC and KDM1A increases apoptotic cell death in vitro. However, whether this combination also increases death of the glioma stem
Ghanshyam Upadhyay et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(22), 8071-8076 (2014-05-21)
Lysine-specific demethylase 1 (LSD1) demethylates nucleosomal histone H3 lysine 4 (H3K4) residues in collaboration with the corepressor CoREST/REST corepressor 1 (Rcor1) and regulates cell fates by epigenetically repressing gene targets. The balanced regulation of this demethylase, if any, is however
Stefano Amente et al.
Oncotarget, 6(16), 14572-14583 (2015-06-13)
The chromatin-modifying enzyme lysine-specific demethylase 1, KDM1A/LSD1 is involved in maintaining the undifferentiated, malignant phenotype of neuroblastoma cells and its overexpression correlated with aggressive disease, poor differentiation and infaust outcome. Here, we show that LSD1 physically binds MYCN both in
Sathiya Pandi Narayanan et al.
Cancer letters, 367(2), 162-172 (2015-08-01)
The histone demethylase KDM1A specifically demethylates lysine residues and its deregulation has been implicated in the initiation and progression of various cancers. However, KDM1A's molecular role and its pathological consequences, and prognostic significance in oral cancer remain less understood. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service