Skip to Content
Merck
All Photos(1)

Key Documents

EHU153641

Sigma-Aldrich

MISSION® esiRNA

targeting human HRH4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTGAATGGGCCAATGATTCTAGTTTCAGAGTCTTGGAAGGATGAAGGTAGTGAATGTGAACCTGGATTTTTTTCGGAATGGTACATCCTTGCCATCACATCATTCTTGGAATTCGTGATCCCAGTCATCTTAGTCGCTTATTTCAACATGAATATTTATTGGAGCCTGTGGAAGCGTGATCATCTCAGTAGGTGCCAAAGCCATCCTGGACTGACTGCTGTCTCTTCCAACATCTGTGGACACTCATTCAGAGGTAGACTATCTTCAAGGAGATCTCTTTCTGCATCGACAGAAGTTCCTGCATCCTTTCATTCAGAGAGACAGAGGAGAAAGAGTAGTCTCATGTTTTCCTCAAGAACCAAGATGAATAGCAATACAATTGCTTCCAAAATGGGTTCCTTCTCCCAATCAGATTCTGTAGCTCTTCACCAAAGGGAACA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angel Jemima Ebenezer et al.
Journal of receptor and signal transduction research, 38(3), 204-212 (2018-06-05)
Mast cell (MC) activation through H4R releases various inflammatory mediators which are associated with allergic asthma. To investigate the siRNA-mediated gene silencing effect of H4R on human mast cells (HMCs) functions and the activation of stress-activated protein kinases (SAPK)/jun amino-terminal
Anne-France Petit-Bertron et al.
PloS one, 4(8), e6504-e6504 (2009-08-08)
The most recently characterized H4 histamine receptor (H4R) is expressed preferentially in the bone marrow, raising the question of its role during hematopoiesis. Here we show that both murine and human progenitor cell populations express this receptor subtype on transcriptional
Angel Jemima Ebenezer et al.
Journal of receptor and signal transduction research, 37(2), 133-140 (2016-07-13)
The histamine H4 receptor functionally expressed on human mast cells and their signaling pathways for the production of IL-13 and RANTES have never been analyzed side by side in a directly comparable manner. Therefore, the aim of the study was
Ya-Xin Zhao et al.
American journal of physiology. Endocrinology and metabolism, 317(6), E1158-E1171 (2019-09-25)
Although many studies have shown that histamine and its signaling regulate energy homeostasis through the central nervous system, their roles in adipose tissues remain poorly understood. Here, we identified that the histamine H4 receptor (HrH4) was highly expressed in adipocytes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service