Skip to Content
Merck
All Photos(1)

Key Documents

EHU117241

Sigma-Aldrich

MISSION® esiRNA

targeting human CDC42

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACTGCAGGGCAAGAGGATTATGACAGATTACGACCGCTGAGTTATCCACAAACAGATGTATTTCTAGTCTGTTTTTCAGTGGTCTCTCCATCTTCATTTGAAAACGTGAAAGAAAAGTGGGTGCCTGAGATAACTCACCACTGTCCAAAGACTCCTTTCTTGCTTGTTGGGACTCAAATTGATCTCAGAGATGACCCCTCTACTATTGAGAAACTTGCCAAGAACAAACAGAAGCCTATCACTCCAGAGACTGCTGAAAAGCTGGCCCGTGACCTGAAGGCTGTCAAGTATGTGGAGTGTTCTGCACTTACACA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arjun P Athreya et al.
Oncotarget, 8(16), 27199-27215 (2017-04-21)
We demonstrate that model-based unsupervised learning can uniquely discriminate single-cell subpopulations by their gene expression distributions, which in turn allow us to identify specific genes for focused functional studies. This method was applied to MDA-MB-231 breast cancer cells treated with
Huanxin Ma et al.
Frontiers in oncology, 10, 438-438 (2020-05-01)
PIWI-interacting RNA (piRNA) is a kind of non-coding single stranded RNA which plays major roles in epigenetic expressions, genome rearrangement, and regulation of gene and protein. Because piRNAs are abnormally expressed in various cancers, they can be used as novel
Xiaolei Liu et al.
Development (Cambridge, England), 145(17) (2018-07-26)
Although major progress in our understanding of the genes and mechanisms that regulate lymphatic vasculature development has been made, we still do not know how lumen formation and maintenance occurs. Here, we identify the Ras-interacting protein Rasip1 as a key
Shayoni Ray et al.
Molecular biology of the cell, 25(16), 2393-2407 (2014-06-27)
Coordinated actin microfilament and microtubule dynamics is required for salivary gland development, although the mechanisms by which they contribute to branching morphogenesis are not defined. Because LIM kinase (LIMK) regulates both actin and microtubule organization, we investigated the role of
Tilman D Rachner et al.
Breast cancer research : BCR, 16(1), R20-R20 (2014-02-18)
Amino-bisphosphonates and statins inhibit the mevalonate pathway, and may exert anti-tumor effects. The Wnt inhibitor dickkopf-1 (DKK-1) promotes osteolytic bone lesions by inhibiting osteoblast functions and has been implicated as an adverse marker in multiple cancers. We assessed the effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service