Skip to Content
Merck
All Photos(1)

Documents

EHU113241

Sigma-Aldrich

MISSION® esiRNA

targeting human G3BP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GAGCCTCAGGAGGAGTCTGAAGAAGAAGTAGAGGAACCTGAAGAAAGACAGCAAACACCTGAGGTGGTACCTGATGATTCTGGAACTTTCTATGATCAGGCAGTTGTCAGTAATGACATGGAAGAACATTTAGAGGAGCCTGTTGCTGAACCAGAGCCTGATCCTGAACCAGAACCAGAACAAGAACCTGTATCTGAAATCCAAGAGGAAAAGCCTGAGCCAGTATTAGAAGAAACTGCCCCTGAGGATGCTCAGAAGAGTTCTTCTCCAGCACCTGCAGACATAGCTCAGACAGTACAGGAAGACTTGAGGACATTTTCTTGGGCATCTGTGACCAGTAAGAATCTTCCACCCAGTGGAGCTGTTCCAGTTACTGGGATACCACCTCATGTTGTTAAAGTACCAGCTTCACAGCCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Dou et al.
American journal of cancer research, 6(11), 2641-2650 (2016-12-03)
RasGAP SH3-domain-Binding Protein 1 (G3BP1) has been implicated in cell growth, migration, and metastasis of some cancers, yet its function in hepatocellular carcinoma (HCC) remains to be explored. In the present study, we reported that G3BP1 was upregulated in HCC
Amr Omer et al.
EMBO reports, 19(5) (2018-03-30)
Cellular senescence is a physiological response by which an organism halts the proliferation of potentially harmful and damaged cells. However, the accumulation of senescent cells over time can become deleterious leading to diseases and physiological decline. Our data reveal a
Allison B Herman et al.
Arteriosclerosis, thrombosis, and vascular biology, 39(10), 2014-2027 (2019-08-30)
Stress granules (SGs) are dynamic cytoplasmic aggregates containing mRNA, RNA-binding proteins, and translation factors that form in response to cellular stress. SGs have been shown to contribute to the pathogenesis of several human diseases, but their role in vascular diseases
Amr Omer et al.
Nature communications, 11(1), 4979-4979 (2020-10-07)
Cellular senescence is a known driver of carcinogenesis and age-related diseases, yet senescence is required for various physiological processes. However, the mechanisms and factors that control the negative effects of senescence while retaining its benefits are still elusive. Here, we
David Pla-Martín et al.
The EMBO journal, 39(9), e102731-e102731 (2020-03-10)
Mitochondria house anabolic and catabolic processes that must be balanced and adjusted to meet cellular demands. The RNA-binding protein CLUH (clustered mitochondria homolog) binds mRNAs of nuclear-encoded mitochondrial proteins and is highly expressed in the liver, where it regulates metabolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service