Skip to Content
Merck
All Photos(1)

Key Documents

EHU105721

Sigma-Aldrich

MISSION® esiRNA

targeting human PCNA

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
R$1,765.00
50 μG
R$3,150.00

R$1,765.00


Estimated to ship onMay 16, 2025



Select a Size

Change View
20 μG
R$1,765.00
50 μG
R$3,150.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

R$1,765.00


Estimated to ship onMay 16, 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCGTGAACCTCACCAGTATGTCCAAAATACTAAAATGCGCCGGCAATGAAGATATCATTACACTAAGGGCCGAAGATAACGCGGATACCTTGGCGCTAGTATTTGAAGCACCAAACCAGGAGAAAGTTTCAGACTATGAAATGAAGTTGATGGATTTAGATGTTGAACAACTTGGAATTCCAGAACAGGAGTACAGCTGTGTAGTAAAGATGCCTTCTGGTGAATTTGCACGTATATGCCGAGATCTCAGCCATATTGGAGATGCTGTTGTAATTTCCTGTGCAAAAGACGGAGTGAAATTTTCTGCAAGTGGAGAACTTGGAAATGGAAACATTAAATTGTCACAGACAAGTAATGTCGATAAAGAGGAGGAAGCTGTTACCATAGAGATGAATGAACCAGTTCAACTAACTTTTGCACTGAGGTACCTGAACTTCTTTACAAAAGCCACTCCACTCTCTTCAACGGTGACACTCAGTATGTCTGCAGATGTACCCCTTGTTGTAGAGTATAAAATTGCGGATATGGGACA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy Kwan et al.
Molecular cancer therapeutics, 20(3), 589-601 (2020-12-11)
Oncolytic viruses (OV) have been shown to activate the antitumor functions of specific immune cells like T cells. Here, we show OV can also reprogram tumor-associated macrophage (TAM) to a less immunosuppressive phenotype. Syngeneic, immunocompetent mouse models of primary breast
Keiichiro Sakuma et al.
Cancer science, 109(8), 2458-2468 (2018-06-06)
Heterogeneous nuclear ribonucleoprotein L-like (HNRNPLL), an RNA-binding protein that regulates alternative splicing of pre-mRNA, has been shown to regulate differentiation of lymphocytes, as well as metastasis of colorectal cancer cells. Here, we show that HNRNPLL promotes cell cycle progression and
N Kanu et al.
Oncogene, 35(30), 4009-4019 (2015-11-10)
The DNA replication machinery invariably encounters obstacles that slow replication fork progression, and threaten to prevent complete replication and faithful segregation of sister chromatids. The resulting replication stress activates ATR, the major kinase involved in resolving impaired DNA replication. In
Wanwan Jia et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 32(7), 4031-4042 (2018-02-27)
Rheumatoid arthritis (RA) is an immune-mediated disease with the characteristics of progressive joint destruction, deformity, and disability. Epigenetic changes have been implicated in the development of some autoimmune disorders, resulting in an alteration of gene transcription. Here, we investigated how
Joachim Garbrecht et al.
Nucleus (Austin, Tex.), 9(1), 474-491 (2018-09-13)
Fluorescence microscopy in combination with the induction of localized DNA damage using focused light beams has played a major role in the study of protein recruitment kinetics to DNA damage sites in recent years. Currently published methods are dedicated to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service