Skip to Content
Merck
All Photos(1)

Key Documents

EHU088591

Sigma-Aldrich

MISSION® esiRNA

targeting human NR3C2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CATTCATGTTCAGGCACCTCTTTTAAAGGGAATCCAACAGTAAACCCGTTTCCATTTATGGATGGCTCGTATTTTTCCTTTATGGATGATAAAGACTATTATTCCCTATCAGGAATTTTAGGACCACCTGTGCCCGGCTTTGATGGTAACTGTGAAGGCAGCGGATTCCCAGTGGGTATTAAACAAGAACCAGATGATGGGAGCTATTACCCAGAGGCCAGCATCCCTTCCTCTGCTATTGTTGGGGTGAATTCAGGTGGACAGTCCTTCCACTACAGGATTGGTGCTCAAGGTACAATATCTTTATCACGATCGGCTAGAGACCAATCTTTCCAACACCTGAGTTCCTTTCCTCCTGTCAATACTTTAGTGGAGTCATGGAAATCACACGGCGACCTGTCGTCTAGAAGAAGTGATGGGTATCCGGTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moshe Shashar et al.
Nitric oxide : biology and chemistry, 80, 24-31 (2018-07-30)
Blockade of the mineralocorticoid receptor (MCR) has been shown to improve endothelial function far beyond blood pressure control. In the current studies we have looked at the effect of MCR antagonists on cationic amino acid transporter-1 (CAT-1), a major modulator
Kunihiro Hayakawa et al.
International journal of molecular sciences, 20(4) (2019-02-17)
MicroRNA (miRNA) is small RNA of 20 to 22 nucleotides in length and is stably present in plasma. Regulating the expression of miRNA taken into cells has been suggested as a general therapeutic approach. We identified the novel anti-inflammatory miRNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service