Skip to Content
Merck
All Photos(1)

Key Documents

EHU065551

Sigma-Aldrich

MISSION® esiRNA

targeting human ATP11C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAGAAAGAGCGAGAGACCTTGAAGGTTTTAAAAATGTTCACCGACTTCCTATCATTTATGGTTCTATTCAACTTTATCATTCCTGTCTCCATGTACGTCACAGTAGAAATGCAGAAATTCTTGGGCTCCTTCTTCATCTCATGGGATAAGGACTTTTATGATGAAGAAATTAATGAAGGAGCCCTGGTTAACACATCAGACCTTAATGAAGAACTTGGTCAGGTGGATTATGTATTTACAGATAAGACTGGAACACTCACTGAAAACAGCATGGAATTCATTGAATGCTGCATAGATGGCCACAAATATAAAGGTGTAACTCAAGAGGTTGATGGATTATCTCAAACTGATGGAACTTTAACATATTTTGACAAAGTAGATAAGAATCGAGAAGAGCTGTTTCTACGTGCCTTGTGTTTATGTCATACTGTAGAAATCAAAACAAACGATGCTGTTGATGGAGCTA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra Ghiani et al.
PloS one, 11(5), e0155803-e0155803 (2016-05-18)
Tomato (Solanum lycopersicum) is one of the most extensively consumed vegetables but, unfortunately, it is also able to induce allergic reactions. In the past, it has been shown that the choice of tomato cultivar significantly influenced the allergic reaction of
Donatien Kamdem Toukam et al.
PloS one, 7(5), e38068-e38068 (2012-06-06)
Although human immunodeficiency type 1 (HIV-1) infection induces strong antibody responses to the viral envelope glycoprotein (Env) only a few of these antibodies possess the capacity to neutralize a broad range of strains. The induction of such antibodies represents an
Theo Rispens et al.
PloS one, 8(2), e55566-e55566 (2013-02-08)
IgE antibodies to gal-α-1,3-gal-β-1,4-GlcNAc (α-gal) can mediate a novel form of delayed anaphylaxis to red meat. Although IgG antibodies to α-gal (anti-α-gal or anti-Gal) are widely expressed in humans, IgE anti-α-gal is not. We explored the relationship between the IgG
Pallara Janardhanan Wills et al.
PloS one, 11(4), e0152787-e0152787 (2016-04-14)
Lepidopterism is a disease caused by the urticating scales and toxic fluids of adult moths, butterflies or its caterpillars. The resulting cutaneous eruptions and systemic problems progress to clinical complications sometimes leading to death. High incidence of fever epidemics were
Simone Mader et al.
Journal of neuroinflammation, 8, 184-184 (2011-12-30)
Serum autoantibodies against the water channel aquaporin-4 (AQP4) are important diagnostic biomarkers and pathogenic factors for neuromyelitis optica (NMO). However, AQP4-IgG are absent in 5-40% of all NMO patients and the target of the autoimmune response in these patients is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service