Skip to Content
Merck
All Photos(1)

Key Documents

EHU064011

Sigma-Aldrich

MISSION® esiRNA

targeting human PSAT1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
R$1,765.00
50 μG
R$3,150.00

R$1,765.00


Check Cart for Availability


Select a Size

Change View
20 μG
R$1,765.00
50 μG
R$3,150.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

R$1,765.00


Check Cart for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGTAAAGCTGGTGGGACCTAATGTCACCTTAATTCTGACTTGAACTGGAAGCATTTTAAGAAATCTTGTTGCTTTTCTAACAAATTCCCGCGTATTTTGCCTTTGCTGCTACTTTTTCTAGTTAGATTTCAAACTTGCCTGTGGACTTAATAATGCAAGTTGCGATTAATTATTTCTGGAGTCATGGGAACACACAGCACAGAGGGTAGGGGGGCCCTCTAGGTGCTGAATCTACACATCTGTGGGGTCTCCTGGGTTCAGCGGCTGTTGATTCAAGGTCAACATTGACCATTGGAGGAGTGGTTTAAGAGTGCCAGGCGAAGGGCAAACTGTAGATCGATCTTTATGCTGTTATTACAGGAGAAGTGACATACTTTATATATGTTTATATTAGCAAGGTCTGTTTTTAATACCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yiqun Zhang et al.
OncoTargets and therapy, 13, 5443-5453 (2020-07-02)
A growing number of studies have found that the serine-glycine biosynthesis pathway is highly activated for biosynthesis in cancer progression and metastasis. Phosphoserine aminotransferase 1 (PSAT1) catalyzes the second step of the serine-glycine biosynthesis pathway; the effects and mechanism of
Stephanie Metcalf et al.
Clinical & experimental metastasis, 37(1), 187-197 (2019-10-21)
Breast cancer is the second leading cause of cancer-related deaths among women and 90% of these mortalities can be attributed to progression to metastatic disease. In particular, triple negative breast cancer (TNBC) is extremely aggressive and frequently metastasizes to multiple
Nadia Vié et al.
Molecular cancer, 7, 14-14 (2008-01-29)
Colorectal cancer (CRC) is one of the most common causes of cancer death throughout the world. In this work our aim was to study the role of the phosphoserine aminotransferase PSAT1 in colorectal cancer development. We first observed that PSAT1
J Patrick Murphy et al.
Cell reports, 24(9), 2381-2391 (2018-08-30)
NAD+ is a key metabolic redox cofactor that is regenerated from nicotinamide through the NAD+ salvage pathway. Here, we find that inhibiting the NAD+ salvage pathway depletes serine biosynthesis from glucose by impeding the NAD+-dependent protein, 3-phosphoglycerate dehydrogenase (PHGDH). Importantly, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service