Skip to Content
Merck
All Photos(1)

Key Documents

EHU062201

Sigma-Aldrich

MISSION® esiRNA

targeting human EED

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
R$1,765.00
50 μG
R$3,150.00

R$1,765.00


Estimated to ship onMay 07, 2025



Select a Size

Change View
20 μG
R$1,765.00
50 μG
R$3,150.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

R$1,765.00


Estimated to ship onMay 07, 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGAAGTCAAAGAAATGCAAATATTCTTTCAAATGTGTAAATAGTCTCAAGGAAGATCATAACCAACCATTGTTTGGAGTTCAGTTTAACTGGCACAGTAAAGAAGGAGATCCATTAGTGTTTGCAACTGTAGGAAGCAACAGAGTTACCTTGTATGAATGTCATTCACAAGGAGAAATCCGGTTGTTGCAATCTTACGTGGATGCTGATGCTGATGAAAACTTTTACACTTGTGCATGGACCTATGATAGCAATACGAGCCATCCTCTGCTGGCTGTAGCTGGATCTAGAGGCATAATTAGGATAATAAATCCTATAACAATGCAGTGTATAAAGCACTATGTTGGCCATGGAAATGCTATCAATGAGCTGAAATTCCATCCAAGAGATCCAAATCTTCTCCTGTCAGTAAGTAAAGATCATGCTTTACGATTATGGAATATCCAGACGGACACTCTGGTGGCAATATTTGGAGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bing He et al.
Oncotarget, 8(61), 103919-103930 (2017-12-22)
The miRNAs play important regulating roles in the pathogenesis of hepatocellular carcinoma (HCC). To uncover key regulating miRNAs in HCC that were neglected by traditional analyzing methods of transcriptomics data, we proposed a novel molecular-network-based omics' (MNBO) method. With this
Houliang Deng et al.
Scientific reports, 9(1), 197-197 (2019-01-19)
Chromobox 6 (CBX6) is a subunit of Polycomb Repressive Complex 1 (PRC1) that mediates epigenetic gene repression and acts as an oncogene or tumor suppressor in a cancer type-dependent manner. The specific function of CBX6 in breast cancer is currently
Qipeng Liu et al.
International journal of cancer, 145(2), 415-426 (2019-01-11)
Polycomb group proteins are important epigenetic regulators for cell proliferation and differentiation, organ development, as well as initiation and progression of lethal diseases, including cancer. Upregulated Polycomb group proteins, including Enhancer of zeste homolog 2 (EZH2), promote proliferation, migration, invasion
Weisi Liu et al.
The Journal of biological chemistry, 290(47), 28489-28501 (2015-10-08)
Our previous studies identified the oncogenic role of p21-activated kinase 1 (PAK1) in hepatocellular carcinoma (HCC) and renal cell carcinoma (RCC). Contrarily, PAK6 was found to predict a favorable prognosis in RCC patients. Nevertheless, the ambiguous tumor suppressive function of
Seong Won Lee et al.
Developmental cell, 46(1), 73-84 (2018-07-06)
The ability to convert human somatic cells efficiently to neurons facilitates the utility of patient-derived neurons for studying neurological disorders. As such, ectopic expression of neuronal microRNAs (miRNAs), miR-9/9∗ and miR-124 (miR-9/9∗-124) in adult human fibroblasts has been found to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service