Skip to Content
Merck
All Photos(1)

Key Documents

EHU045261

Sigma-Aldrich

MISSION® esiRNA

targeting human SPRY2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGAGCTTAAGCCACTGAGCAAGGAAGATTTGGGCCTGCACGCCTACAGGTGTGAGGACTGTGGCAAGTGCAAATGTAAGGAGTGCACCTACCCAAGGCCTCTGCCATCAGACTGGATCTGCGACAAGCAGTGCCTTTGCTCGGCCCAGAACGTGATTGACTATGGGACTTGTGTATGCTGTGTGAAAGGTCTCTTCTATCACTGTTCTAATGATGATGAGGACAACTGTGCTGACAACCCATGTTCTTGCAGCCAGTCTCACTGTTGTACACGATGGTCAGCCATGGGTGTCATGTCCCTCTTTTTGCCTTGTTTATGGTGTTACCTTCCAGCCAAGGGTTGCCTTAAATTGTGCCAGGGGTGTTATGACCGGGTTAACAGGCCTGGTTGCCGCTGTAAAAACTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-Ho Ahn et al.
Biomolecules & therapeutics, 23(4), 320-326 (2015-07-15)
The clinical benefits of oncogenic BRAF inhibitor therapies are limited by the emergence of drug resistance. In this study, we investigated the role of a negative regulator of the MAPK pathway, Spry2, in acquired resistance using BRAF inhibitor-resistant derivatives of
Yan Liu et al.
Molecular medicine reports, 23(5) (2021-03-25)
Age-related cataract (ARC) is the primary cause of blindness worldwide. Abnormal expression of microRNAs (miRNAs/miRs) has been reported to be associated with multiple diseases, including ARC. However, the potential role of miR-124 in ARC remains unclear. The present study used
Evan K Day et al.
Cell reports, 30(10), 3383-3396 (2020-03-12)
SPRY2 is a purported tumor suppressor in certain cancers that promotes tumor growth and resistance to receptor tyrosine kinase inhibitors in glioblastoma. Here, we identify a SPRY2-dependent bypass signaling mechanism in glioblastoma that drives resistance to EGFR and MET inhibition.
Tatiana Lopatina et al.
Oncogenesis, 9(10), 90-90 (2020-10-12)
The lack of approved targeted therapies highlights the need for new treatments for triple-negative breast cancer (TNBC) patients. Interleukin-3 (IL-3) acts as an autocrine factor for tumor-endothelial cells (TEC), and exerts pro-angiogenic paracrine action via extracellular vesicles (EVs). IL-3Rα blockade

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service