Skip to Content
Merck
All Photos(1)

Documents

EHU039111

Sigma-Aldrich

MISSION® esiRNA

targeting human LPAR4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTTGAAGGCTTCTCCAAACGTGTCTGGAAGACTTATTTATCCAAGATCACAATATTTATTGAAGTTGTTGGGTTTATCATTCCTCTAATATTGAATGTCTCTTGCTCTTCTGTGGTGCTGAGAACTCTTCGCAAGCCTGCTACTCTGTCTCAAATTGGGACCAATAAGAAAAAAGTACTGAAAATGATCACAGTACATATGGCAGTCTTTGTGGTATGCTTTGTACCCTACAACTCTGTCCTCTTCTTGTATGCCCTGGTGCGCTCCCAAGCTATTACTAATTGCTTTTTGGAAAGATTTGCAAAGATCATGTACCCAATCACCTTGTGCCTTGCAACTCTGAACTGTTGTTTTGACCCTTTCATCTATTACTTCACCCTTGAATCCTTTCAGAAGTCCTTCTACATCAATGCCCACATCAGAATGGAGTCCCTGTTTAAGACTGAAACACCTTTGACCACAAAGCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen Jiang et al.
Journal of cellular biochemistry, 119(8), 6429-6441 (2017-12-15)
The study explores whether miR-139-5p targeting LPAR4 affects epithelial-mesenchymal transition (EMT) and fibrosis in post-menopausal women with interstitial cystitis (IC) via the PI3K/Akt signaling pathway. Bladder tissues of IC and normal bladder tissues were collected. The pathology of bladder tissues
Nobuhiro Kurabayashi et al.
Development (Cambridge, England), 145(17) (2018-09-16)
Newborn neurons in the developing neocortex undergo radial migration, a process that is coupled with their precise passage from multipolar to bipolar shape. The cell-extrinsic signals that govern this transition are, however, poorly understood. Here, we find that lysophosphatidic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service