Skip to Content
Merck
All Photos(1)

Key Documents

EHU019271

Sigma-Aldrich

MISSION® esiRNA

targeting human FABP4

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
R$1,765.00
50 μG
R$3,150.00

R$1,765.00


Estimated to ship onApril 19, 2025



Select a Size

Change View
20 μG
R$1,765.00
50 μG
R$3,150.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

R$1,765.00


Estimated to ship onApril 19, 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGCAGCTTCCTTCTCACCTTGAAGAATAATCCTAGAAAACTCACAAAATGTGTGATGCTTTTGTAGGTACCTGGAAACTTGTCTCCAGTGAAAACTTTGATGATTATATGAAAGAAGTAGGAGTGGGCTTTGCCACCAGGAAAGTGGCTGGCATGGCCAAACCTAACATGATCATCAGTGTGAATGGGGATGTGATCACCATTAAATCTGAAAGTACCTTTAAAAATACTGAGATTTCCTTCATACTGGGCCAGGAATTTGACGAAGTCACTGCAGATGACAGGAAAGTCAAGAGCACCATAACCTTAGATGGGGGTGTCCTGGTACATGTGCAGAAATGGGATGGAAAATCAACCACCATAAAGAGAAAACGAGAGGATGATAAACTGGTGGTGGAATGCGTCATGAAAGGCGTCACTTCCAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Wang et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 85, 272-279 (2016-12-05)
FABP4 is widely expressed in both normal and pathologic tissues. It promotes cell proliferation, survival and migration of endothelial cells, and therefore, angiogenesis. However, the role of FABP4 in hemangioma or hemangioma endothelial cells (HemECs) has not been explored. In
Sanjay Basak et al.
Molecular and cellular biochemistry, 437(1-2), 55-64 (2017-06-18)
Adequate placental angiogenesis is critical for the establishment of the placental circulation and thus for normal feto-placental growth and development. Fatty acid-binding protein-4 (FABP4) plays a pro-angiogenic role in endothelial cells; however, very little information is available in placental first
U Harjes et al.
Oncogene, 36(7), 912-921 (2016-08-30)
Fatty acid binding protein 4 (FABP4) is a fatty acid chaperone, which is induced during adipocyte differentiation. Previously we have shown that FABP4 in endothelial cells is induced by the NOTCH1 signalling pathway, the latter of which is involved in
Mingguo Huang et al.
Oncotarget, 8(67), 111780-111794 (2018-01-18)
Fatty acid binding protein 4 (FABP4) is an abundant protein in adipocytes, and its production is influenced by high-fat diet (HFD) or obesity. The prostate stromal microenvironment induces proinflammatory cytokine production, which is key for the development and progression of
Rebecca F Rogers et al.
Cancer research, 80(8), 1735-1747 (2020-03-13)
Checkpoint kinase 1 (CHK1) is a key mediator of the DNA damage response that regulates cell-cycle progression, DNA damage repair, and DNA replication. Small-molecule CHK1 inhibitors sensitize cancer cells to genotoxic agents and have shown single-agent preclinical activity in cancers

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service