Skip to Content
Merck
All Photos(1)

Key Documents

EHU017091

Sigma-Aldrich

MISSION® esiRNA

targeting human RRM1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCCAATAAAGATCGCCTGAATTCTGCTATTATCTATGACCGAGATTTCTCTTACAATTACTTCGGCTTTAAGACGCTAGAGCGGTCTTATTTGTTGAAGATCAATGGAAAAGTGGCTGAAAGACCACAACATATGTTGATGAGAGTATCTGTTGGGATCCACAAAGAAGACATTGATGCAGCAATTGAAACATATAATCTTCTTTCTGAGAGGTGGTTTACTCATGCTTCGCCCACTCTCTTCAATGCTGGTACCAACCGCCCACAACTTTCTAGCTGTTTTCTTCTGAGTATGAAAGATGACAGCATTGAAGGCATTTATGACACTCTAAAGCAATGTGCATTGATTTCTAAGTCTGCTGGAGGAATTGGTGTTGCTGTGAGTTGTATTCGGGCTACTGGCAGCTACATTGCTGGGACTAATGGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zarah Glad Zimling et al.
Anticancer research, 35(12), 6731-6738 (2015-12-08)
A possible predictive impact of ribonucleotide-reductase subunit-1 (RRM1) on vinorelbine efficacy in non-small cell lung cancer (NSCLC) has been previously reported. The present study aimed to further explore this finding in malignant pleural mesothelioma (MPM). Seventy-one patients with MPM receiving
Zhen Shu et al.
Oncogene, 39(35), 5721-5733 (2020-07-28)
Ribonucleotide reductase (RNR) catalyzes the rate-limiting step of de novo synthesis of deoxyribonucleotide triphosphates (dNTPs) building blocks for DNA synthesis, and is a well-recognized target for cancer therapy. RNR is a heterotetramer consisting of two large RRM1 subunits and two
Ribonucleotide Reductase Catalytic Subunit M1 (RRM1) as a Novel Therapeutic Target in Multiple Myeloma.
Morihiko Sagawa et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 23(17), 5225-5237 (2017-04-27)
Rong Yao et al.
PloS one, 10(5), e0125169-e0125169 (2015-05-07)
Pulmonary fibrosis is one of the most common complications of paraquat (PQ) poisoning, which demands for more effective therapies. Accumulating evidence suggests adiponectin (APN) may be a promising therapy against fibrotic diseases. In the current study, we determine whether the
Jianmei Wu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1006, 167-178 (2015-11-10)
Simultaneous, quantitative determination of intracellular nucleoside triphosphates and other polar metabolites using liquid chromatography with electrospray ionization tandem mass spectrometry (LC-MS/MS) represents a bioanalytic challenge because of charged, highly hydrophilic analytes presented at a large concentration range in a complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service