Skip to Content
Merck
All Photos(4)

Documents

EC500

Sigma-Aldrich

GenElute-E Single Spin Plant DNA Kit

greener alternative

Synonym(s):

GenElute-E Plant DNA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600
NACRES:
NA.52

purified by

(Single-spin negative chromotography)
(Time: 45 minutes or less)

feature

Compatible Application (Suitable for most common downstream applications, including genotyping, PCR, and NGS)
Intended use (For purification of genomic DNA from plant tissue)
Typical/expected yield (Varies by sample. Please reference user guide for more information.)

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

General description

GenElute-E purification kits provide a simple alternative for DNA purification. Compared with traditional methods, GenElute-E offers:
  • Isolation of DNA in under an hour with minimal sample handling
  • Better purity, leading to improved performance in PCR and other applications
  • Significant reduction in plastic and hazardous chemical waste


Traditional silica-based, bind-wash-elute purification kits require multiple wash steps to remove impurities from the spin columns. These steps increase the risk of cross contamination, subject the DNA to centrifugation sheering forces, and introduce chaotropic salts that can carry over into the final sample, inhibiting downstream applications.

GenElute-E kits employ size exclusion negative chromatography to separate large nucleic acid molecules from smaller protein, lipid, and ionic components in cell, tissue, blood, and other samples. Single-spin columns efficiently absorb and retain cellular debris and sample contaminants while allowing nucleic acids to pass through, reducing the number of steps and plastic materials required for purification. This novel method for high-quality purification is made possible by our innovative SmartLyse® Protease, which enables fast and efficient lysis of a wide range of sample types.

Isolate nucleic acid in a fraction of time compared to traditional silica bind-wash-elute procedures. Simply mix sample with lysis components and incubate according to kit protocol, pipette sample onto the spin column, and centrifuge directly into a collection tube. Cellular debris and contaminants remain bound in the column to be discarded, while purified DNA is ready to be used in downstream applications or stored.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is aligned with "Waste prevention" and "Designing safer chemicals". Click here for more information.

Application

GenElute-E Single Spin Plant DNA Kit quickly purifies genomic DNA from various species and types of plant tissues, including leaf, seed, fruit and root material.

The included grinding suspension enables a fast and effective disruption of plant tissue without bead beating. The subsequent lysis step takes place under physiological conditions where the enzyme activity of proteases is generally increased. SmartLyse® Protease contains a unique combination of several active enzymes with an expanded substrate specificity. This eliminates the requirement for extended, overnight incubations, which are required in many conventional protocols.

Purified genomic DNA is eluted in Tris buffer, pH 7.8 and can immediately be used for most common downstream applications, including PCR, genotyping, NGS, and others. Final yield is equal or better than silica-based methods depending on sample type. GenElute-E purified genomic DNA preparations commonly show an A260/280 ratio of around 1.8.

Features and Benefits

Fast and efficient purification: 40-minute lysis and minimal hands-on, preparation stepsHigher quality DNA: negative chromotography-based purification reduces carryover of PCR inhibitors and other contaminents Reduced environmental impact: 55% less plastic waste, as well as a significant reduction in hazardous chemicals

Storage and Stability

Store enzymes at 2-8°C for up to 12 months. All other components should be stored at room temperature.

Other Notes

We believe that green chemistry will contribute to a better tomorrow. With a growing portfolio of greener alternatives, there are now more choices to reduce the ecological impact of your research while still delivering quality and efficacy so your results are not compromised.  GenElute-E purification kits avoid the use of hazardous wash buffers typically employed in silica-based purification kits, significantly reducing chemical waste. The single-spin protocol also limits the number of collection steps, reducing plastic waste by 55% compared to equivalent bind-wash-elute methods. GenElute-E is designated a Greener Alternative Product based on the "Prevention" and "Designing Safer Chemicals" qualifications of The 12 Principles of Green Chemistry.GenElute-E kits also adhere to the principles of SMASH Packaging, our global initiative to improve the sustainability through the reduction of packaging materials, the increased use of sustainable alternatives, and improved recyclability. Packaging is FCS-marked for sustainable forestry practices and produced from greater than 70% recycled content.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC
SmartLyse is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

96.8 °F

Flash Point(C)

36 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

GenElute™-E DNA purification kits ensure accurate DNA quantitation and downstream enzymatic processes.

Related Content

GenElute™-E Single Spin DNA and RNA purification technology enables rapid DNA and RNA extraction and purification from mammalian cells, blood, tissue, and plant tissue samples.

Answers to frequently asked questions related to GenElute™-E single spin DNA and RNA purification and negative chromatography

Reduce plastic waste and eliminate hazardous liquid waste for more sustainable laboratories with GenElute™-E Single Spin DNA and RNA prep kits.

Demo video showing how to purify DNA and RNA GenElute™-E single spin nucleic acid purification kits

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service