Skip to Content
Merck
All Photos(1)

Key Documents

E9156

Sigma-Aldrich

Encainide hydrochloride

≥98% (HPLC), powder

Synonym(s):

(+/-)-4-Methoxy-N-[2-[2-(1-methyl-2-piperidinyl)ethyl]phenyl]benzamide hydrochloride, MJ-9067

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28N2O2 · HCl
CAS Number:
Molecular Weight:
388.93
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

solubility

H2O: >25 mg/mL

originator

Bristol-Myers Squibb

storage temp.

2-8°C

SMILES string

Cl.COc1ccc(cc1)C(=O)Nc2ccccc2CCC3CCCCN3C

InChI

1S/C22H28N2O2.ClH/c1-24-16-6-5-8-19(24)13-10-17-7-3-4-9-21(17)23-22(25)18-11-14-20(26-2)15-12-18;/h3-4,7,9,11-12,14-15,19H,5-6,8,10,13,16H2,1-2H3,(H,23,25);1H

InChI key

OJIIZIWOLTYOBS-UHFFFAOYSA-N

Biochem/physiol Actions

Encainide hydrochloride is a sodium channel blocker and class Ic antiarrhythmic.
Encainide hydrochloride is a sodium channel blocker and class Ic antiarrhythmic. Encainide is a non-chiral antiarrhythmic and benzanilide derivative.

Features and Benefits

This compound was developed by Bristol-Myers Squibb. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Sweeney et al.
Pacing and clinical electrophysiology : PACE, 19(1), 50-60 (1996-01-01)
The mechanisms by which pharmacological agents alter electrical defibrillation are not fully understood. It has been proposed that, in addition to directly stimulating tissue, defibrillation may involve refractory period extension (RPE) produced by the shock. Accordingly, pharmacological agents might modulate
C L Case et al.
Biology of the neonate, 66(6), 330-338 (1994-01-01)
With clinical data suggesting that neonates may be more prone to developing electrophysiologic side effects from encainide, this study investigates the in vitro developmental electrophysiologic effects of encainide and its major metabolites on the action potential parameters of the canine
H R Lu et al.
Journal of cardiovascular pharmacology, 26(1), 132-136 (1995-07-01)
We compared the effects of class I-IV antiarrhythmic agents on the ventricular fibrillation threshold (VFT) induced by electrical stimulation directly on the myocardium in anesthetized, open-chest guinea pigs. VFT was assessed by determining the intensity (mA) of electrical current required
R W Peters et al.
Journal of the American College of Cardiology, 23(2), 283-289 (1994-02-01)
The purpose of this study was to assess the effect of antiarrhythmic drugs on the timing of arrhythmic death. Sudden cardiac death remains a problem of epidemic proportions. Delineating its pathophysiology is an important step in devising preventive measures. Previous
A A Wallace et al.
Journal of cardiovascular pharmacology, 21(3), 397-404 (1993-03-01)
The antiarrhythmic efficacy and proarrhythmic potential of the class IC antiarrhythmic agent encainide were assessed in subacute and chronic postinfarction canine models, respectively. In conscious dogs with spontaneous premature ventricular complexes (PVCs) at 48 h after anterior myocardial infarction (MI)

Articles

Voltage-gated sodium channels are present in most excitable cell membranes and play an important role in generating action potentials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service