Skip to Content
Merck
All Photos(1)

Key Documents

E1904

Sigma-Aldrich

Ethyl-3,4-dephostatin

>99% (HPLC), solid

Synonym(s):

3,4-Dihydroxy-N-ethyl-N-nitrosoaniline

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$1,565.00

R$1,565.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
1 MG
R$1,565.00

About This Item

Empirical Formula (Hill Notation):
C8H10N2O3
CAS Number:
Molecular Weight:
182.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$1,565.00


Estimated to ship onMay 28, 2025


Assay

>99% (HPLC)

form

solid

color

brown

solubility

DMSO: soluble 22 mg/mL
H2O: soluble 8 mg/mL

storage temp.

2-8°C

SMILES string

CCN(N=O)c1ccc(O)c(O)c1

InChI

1S/C8H10N2O3/c1-2-10(9-13)6-3-4-7(11)8(12)5-6/h3-5,11-12H,2H2,1H3

InChI key

ZTXUSFPBLYQDDN-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

Stable analog of dephostatin. Selective inhibitor of protein tyrosine phosphatase 1B and SHPTP-1. Does not inhibit CD45-associated phosphatases.

Features and Benefits

This compound is featured on the Phosphoprotein Phosphatases (Tyrosine) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Store under nitrogen

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Kim et al.
Cell death & disease, 4, e588-e588 (2013-04-13)
Hypoxia enhances the proliferation and migration of adipose-derived stem cells (ASCs) via the generation of reactive oxygen species (ROS). Therefore, this study primarily investigated whether or not ROS generation could regulate microRNA-210 (miR-210) expression, and increase proliferation/migration of ASCs. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service