Skip to Content
Merck
All Photos(1)

Documents

E1283

Sigma-Aldrich

ERK2, active, GST tagged human

recombinant, expressed in E. coli, PRECISIO® Kinase, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ERT1, MAPK1, MAPK2, P41mapk, P42MAPK, PRKM1, PRKM2, p40, p47

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

613-829 nmol/min·mg

mol wt

~68 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAPK1(5594)

Biochem/physiol Actions

ERK2 is a protein serine/threonine kinase that is a member of the extracellular signal-regulated kinases (ERKs) which are activated in response to numerous growth factors and cytokines. Activation of ERK2 requires both tyrosine and threonine phosphorylation that is mediated by MEK. ERK2 is ubiquitously distributed in tissues with the highest expression in heart, brain and spinal cord. Activated ERK2 translocates into the nucleus where it phosphorylates various transcription factors (e.g., Elk-1, c-Myc, c-Jun, c-Fos, and C/EBP beta).

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baochi Ou et al.
Cell death & disease, 12(8), 737-737 (2021-07-28)
Neutrophils are significant compositions of solid tumors and exert distinct functions in different types of tumors. However, the precise role of neutrophils in the progression of breast cancer (BC) is presently unclear. In this study, by investigating the single-cell RNA
T G Boulton et al.
Biochemistry, 30(1), 278-286 (1991-01-08)
In rat 1 fibroblasts, insulin has little or no stimulatory effect on the activities of either MAP2 protein kinase or ribosomal protein S6 kinase. In contrast, in rat 1 cells that overexpress the normal human insulin receptor (rat 1 HIRc

Articles

Lipid Induced Insulin Resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service