Skip to Content
Merck
All Photos(1)

Key Documents

D9636

Sigma-Aldrich

10-(2′,4′-Dinitrophenylazo)-9-phenanthrol

Synonym(s):

DNAF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12N4O5
CAS Number:
Molecular Weight:
388.33
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

solubility

methylene chloride: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1c(\N=N\c2ccc(cc2N(=O)=O)N(=O)=O)c3ccccc3c4ccccc14

InChI

1S/C20H12N4O5/c25-20-16-8-4-2-6-14(16)13-5-1-3-7-15(13)19(20)22-21-17-10-9-12(23(26)27)11-18(17)24(28)29/h1-11,25H/b22-21+

InChI key

WIWQKZWDFCJGQY-QURGRASLSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service