D9625
2′-Deoxyguanosine 5′-monophosphate
≥97%
Synonym(s):
5′-Deoxyguanylic acid, dGMP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H14N5O7P
CAS Number:
Molecular Weight:
347.22
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Recommended Products
Assay
≥97%
form
solid
storage temp.
−20°C
SMILES string
NC1=NC(=O)c2ncn(C3CC(O)C(COP(O)(O)=O)O3)c2N1
InChI
1S/C10H14N5O7P/c11-10-13-8-7(9(17)14-10)12-3-15(8)6-1-4(16)5(22-6)2-21-23(18,19)20/h3-6,16H,1-2H2,(H2,18,19,20)(H3,11,13,14,17)
InChI key
LTFMZDNNPPEQNG-UHFFFAOYSA-N
comparable product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Paolo Mariani et al.
The journal of physical chemistry. B, 113(22), 7934-7944 (2009-05-14)
To solve details of the self-assembling process of guanosine in diluted aqueous solution and to derive a thermodynamical model for quadruplex formation, the structural behavior of deoxyguanosine 5'-monophosphate has been analyzed by in-solution small angle X-ray scattering. The experiments have
Takahito Ohshiro et al.
Micromachines, 11(11) (2020-11-05)
We utilized electrophoresis to control the fluidity of sample biomolecules in sample aqueous solutions inside the nanochannel for single-molecule detection by using a nanochannel-integrated nanogap electrode, which is composed of a nano-gap sensing electrode, nanochannel, and tapered focusing channel. In
Martin Haeubl et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 14(7), 1037-1052 (2009-05-28)
The novel gold porphyrin complex (5,10,15-tris(N-methylpyridinium-4-yl)-20-(1-pyrenyl)-porphyrinato)gold(III) chloride, [Au(III)(TMPy3Pyr1P)]Cl4, was prepared and characterized by optical spectroscopy, high-resolution nuclear magnetic resonance (NMR), and electrospray mass spectrometry. This cationic multichromophore compound exhibits excellent water solubility and does not form aggregates under physiological conditions.
Mingfeng Li et al.
PloS one, 8(1), e54420-e54420 (2013-02-01)
Free extracellular DNA provides nutrition to bacteria and promotes bacterial evolution by inducing excessive mutagenesis of the genome. To understand the influence of extracellular DNA fragments on D. radiodurans, we investigated cell growth and survival after extracellular DNA or dNMPs
Mariko Higuchi et al.
Journal of structural biology, 173(1), 20-28 (2010-10-05)
MutT distinguishes substrate 8-oxo-dGTP from dGTP and also 8-oxo-dGMP from dGMP despite small differences of chemical structures between them. In this paper we show by the method of molecular dynamics simulation that the transition between conformational substates of MutT is
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service