Skip to Content
Merck
All Photos(1)

Key Documents

D9321

Sigma-Aldrich

D-dimer, Human

from human plasma, buffered aqueous solution

Synonym(s):

Fibrin degradation fragment, Fragment D-dimer

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$1,315.00
1 MG
R$4,157.00

R$1,315.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
R$1,315.00
1 MG
R$4,157.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.61

R$1,315.00


Check Cart for Availability

Request a Bulk Order

biological source

human plasma

form

buffered aqueous solution

mol wt

Mw ~200 kDa

shipped in

wet ice

storage temp.

−20°C

General description

The human D-dimer(s) protein is an indicator of a thrombosis event. This protein is the degradation fragment of fibrin mesh that has been stabilized by Factor XIII. This factor crosslinks the E-element to two D-elements. As a result, this crosslink is the final step in the generation of a thrombus.

Application

The human D-dimer protein is a blood test performed in the medical laboratory to diagnose thrombosis. While a negative human D-dimer result rules out thrombosis, a positive test result can indicate thrombosis or other underlying potential health problems. The presence of D-dimer can idicate the existance of a deep venous thrombosis (DVT) or pulomonary embolism (PE).

Physical form

The human D-dimer protein provided in liquid form constisting of 0.05 M sodium phosphate, pH 7.5, 0.15 M sodium chloride and 0.09% sodium azide.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the level of fragment D-dimer in a healthy individual?

    1 answer
    1. The normal level of fragment D-dimer in a healthy individual is reported to be less than 0.37 ug/mL in plasma.Lawler, C.M., et. al, Blood, 76, 1341-1348

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service