D8283
N-(7-Dimethylamino-4-methyl-3-coumarinyl)maleimide
Synonym(s):
DACM-3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H14N2O4
CAS Number:
Molecular Weight:
298.29
Beilstein:
1545346
EC Number:
MDL number:
UNSPSC Code:
12352200
mp
218-220 °C (lit.)
storage temp.
2-8°C
SMILES string
CN(C)c1ccc2C(C)=C(N3C(=O)C=CC3=O)C(=O)Oc2c1
InChI
1S/C16H14N2O4/c1-9-11-5-4-10(17(2)3)8-12(11)22-16(21)15(9)18-13(19)6-7-14(18)20/h4-8H,1-3H3
InChI key
ADEORFBTPGKHRP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service