Skip to Content
Merck
All Photos(1)

Documents

D7924

SAFC

3-(Dimethylamino)propionitrile

liquid

Synonym(s):

DMAPN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2CN
CAS Number:
Molecular Weight:
98.15
Beilstein:
773779
EC Number:
MDL number:
UNSPSC Code:
12352200

form

liquid

color

yellow

refractive index

n20/D 1.426 (lit.)

bp

171 °C/750 mmHg (lit.)

mp

−43 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C)CCC#N

InChI

1S/C5H10N2/c1-7(2)5-3-4-6/h3,5H2,1-2H3

InChI key

MTPJEFOSTIKRSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst similar to TEMED used to form oxygen free radicals from ammonium persulfate. Free radicals cause acrylamide and bis-acrylamide to polymerize.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service