Skip to Content
Merck
All Photos(1)

Key Documents

D7052

Sigma-Aldrich

Dipeptidyl Peptidase IV from porcine kidney

lyophilized powder, ≥10 units/mg protein (Bradford)

Synonym(s):

Dipeptidyl aminopeptidase IV

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

≥10 units/mg protein (Bradford)

mol wt

124 kDa

packaging

vial of ≥0.75 unit

solubility

0.1 M Tris-HCl, pH 8.0: soluble 1 vial/mL, clear, colorless

UniProt accession no.

storage temp.

−20°C

Gene Information

Specificity

DPPIV has a post-proline dipeptidyl aminopeptidase activity that hydrolyzes N-terminal dipeptides from the unsubstituted N-terminus of peptides with the sequence of X-Pro-Z and X-Ala-Z. Where X is a nonspecific residue at the N terminus and Z cannot be proline or hydroxyproline.

Application

Dipeptidyl peptidase IV from porcine kidney has been used in a study to investigate engraftment of donor cells following hepatocyte transplantation. Dipeptidyl peptidase IV from porcine kidney has also been used in a study to investigate the metabolism of glucagon-like peptide-2.
The enzyme from sigma has been used in the determination of DPP-IV inhibitory activity of peptides in porcine skin gelatin hydrolysates.

Biochem/physiol Actions

The pH optimum for the protease is 7.8-8.0. Dipeptidyl peptidase does not require activators. It is inhibited by diisopropyl fluorophosphate, phenylmethanesulfonyl fluoride, and diethyl p-nitrophenyl phosphate.
Native DPPIV is a ubiquitous type II transmembrane glycoprotein and a serine protease of the S9 prolyl-oligopeptidase family. In vivo, it is synthesized with a signal peptide, which functions as the membrane anchoring domain. There is an 88% sequence homology between the human and porcine kidney enzymes. Both exist as homodimers with a subunit molecular weight of ~30 kDa. The high mannose 100 kDa DPPIV precursor is processed in the Golgi to yield a 124 kDa heavily N-and O-linked mature glycoprotein. It is then sorted to the apical membrane through the concerted action of both N- and O-linked glycans and its association with lipid microdomains. The porcine enzyme contains 18.3% carbohydrates, which the glycan composition is 0.9% fucose, 3.4% mannose, 5.1% galactose, 8.2% glucosamine, and 0.7% sialic acid. DPPIV is highly expressed on endothelial cells, epithelial cells, and lymphocytes. It is also present in plasma in its soluble form.

Unit Definition

One unit will produce 1.0 μmole of p-nitroaniline from Gly-L-Pro p-nitroanilide per min in 100 mM Tris-HCl at pH 8.0 at 37 °C.

Physical form

Lyophilized powder containing Tris buffer salts

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dipeptidyl Peptidase-IV Inhibitory Activity of Peptides in Porcine Skin Gelatin Hydrolysates.
Hsu, Kuo-Chiang, et al.
Bioactive Food Peptides in Health and Disease, 205-205 (2013)
Kosho Yamanouchi et al.
Hepatology (Baltimore, Md.), 49(1), 258-267 (2008-11-13)
Engraftment of donor hepatocytes is a critical step that determines the success of hepatocyte transplantation. Rapid and efficient integration of donor cells would enable prompt liver repopulation of these cells in response to selective proliferative stimuli offered by a preparative
A J Kenny et al.
The Biochemical journal, 157(1), 169-182 (1976-07-01)
Dipeptidyl peptidase IV, an enzyme that releases dipeptides from substrates with N-terminal sequences of the forms X-Pro-Y or X-Ala-Y, was purified 300-fold from pig kidney cortex. The kidney is the main source of the enzyme, where it is one of
K M Fukasawa et al.
Biochimica et biophysica acta, 657(1), 179-189 (1981-01-15)
Dipeptidyl peptidase IV (dipeptidylpeptide hydrolase, EC 3.4.14.-) has been purified from the microsomal fraction of pig liver, using an immunoaffinity chromatography, and its properties compared with those of the enzyme purified from pig kidney. The amino acid compositions of both
Maegan L Capitano et al.
The Journal of clinical investigation, 129(6), 2555-2570 (2019-05-21)
The nuclear protein DEK is an endogenous DNA-binding chromatin factor regulating hematopoiesis. DEK is one of only 2 known secreted nuclear chromatin factors, but whether and how extracellular DEK regulates hematopoiesis is not known. We demonstrated that extracellular DEK greatly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service