Skip to Content
Merck
All Photos(1)

Key Documents

D6940

Sigma-Aldrich

R-(–)-Desmethyldeprenyl hydrochloride

>98% (HPLC), solid

Synonym(s):

(R)-N-(1-Phenylpropan-2-yl)prop-2-yn-1-amine hydrochloride, N-Desmethylselegiline hydrochloride, L-Nordeprenyl hydrochloride, Norselegiline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N · HCl
CAS Number:
Molecular Weight:
209.72
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

>98% (HPLC)

form

solid

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

off-white

solubility

H2O: soluble >20 mg/mL

application(s)

forensics and toxicology
veterinary

SMILES string

C[C@@H](NCC#C)CC1=CC=CC=C1.Cl

InChI

1S/C12H15N.ClH/c1-3-9-13-11(2)10-12-7-5-4-6-8-12;/h1,4-8,11,13H,9-10H2,2H3;1H/t11-;/m1./s1

InChI key

KJZZTCSJZCYCQS-RFVHGSKJSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Neuroprotectant; anti-apoptotic.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service