Skip to Content
Merck
All Photos(1)

Key Documents

D6567

Sigma-Aldrich

DRF 2519

≥98% (HPLC), solid

Synonym(s):

5-[[4-[2-(4-Oxo-2H-1,3-benzoxazin3(4H)-yl)ethoxy]phenyl]methyl2,4-thiazolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18N2O5S
Molecular Weight:
398.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

storage condition

under inert gas

color

white

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

O=C1NC(=O)C(Cc2ccc(OCCN3COc4ccccc4C3=O)cc2)S1

Biochem/physiol Actions

DRF-2519 is a dual PPARα/γ activator. DRF-2519 has better reduction of plasma insulin, triglyceride, and free fatty acid levels than those treated with rosiglitazone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service