Skip to Content
Merck
All Photos(1)

Key Documents

D4267

Sigma-Aldrich

N,N-Dimethyl-p-phenylenediamine hemioxalate salt

Synonym(s):

4-(Dimethylamino)aniline hemioxalate salt, 4-Amino-N,N-dimethylaniline hemioxalate salt, DMPPDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H12N2 · 1/2C2H2O4
CAS Number:
Molecular Weight:
181.21
EC Number:
MDL number:
UNSPSC Code:
12352100

storage temp.

2-8°C

SMILES string

OC(=O)C(O)=O.CN(C)c1ccc(N)cc1.CN(C)c2ccc(N)cc2

Caution

Darkens readily when exposed to air.

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service