Skip to Content
Merck
All Photos(2)

Key Documents

D2945

Sigma-Aldrich

Anti-DCDC2 antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DCDC2A, Anti-Doublecortin domain containing 2, Anti-RU2, Anti-RU2S

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~55 kDa
antigen ~70 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

western blot: 1-2 μg/mL using HEK-293T cell lysate expressing human DCDC2

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DCDC2(51473)

General description

DCDC2 belongs to the doublecortin (DCX) superfamily and contains two doublecortin domains. DCDC2 is expressed in fetal and adult CNS.

Application

Anti-DCDC2 antibody produced in rabbit is suitable for western blot analysis at a working concentration of 1-2μg/mL using a HEK-293T cell lysate expressing human DCDC2.

Biochem/physiol Actions

DCDC2 is one of the dyslexia susceptibility genes involved in neuronal migration and other developmental processes. The DCX domain of DCDC2 binds tubulin and enhances microtubule polymerization. Several SNPs within the DCDC2 gene have been associated with dyslexia. It modulates neuronal development in the brain.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Schueler et al.
American journal of human genetics, 96(1), 81-92 (2015-01-06)
Nephronophthisis-related ciliopathies (NPHP-RC) are recessive diseases characterized by renal dysplasia or degeneration. We here identify mutations of DCDC2 as causing a renal-hepatic ciliopathy. DCDC2 localizes to the ciliary axoneme and to mitotic spindle fibers in a cell-cycle-dependent manner. Knockdown of
Johannes Schumacher et al.
American journal of human genetics, 78(1), 52-62 (2005-12-31)
We searched for linkage disequilibrium (LD) in 137 triads with dyslexia, using markers that span the most-replicated dyslexia susceptibility region on 6p21-p22, and found association between the disease and markers within the VMP/DCDC2/KAAG1 locus. Detailed refinement of the LD region
Lauren M McGrath et al.
Trends in molecular medicine, 12(7), 333-341 (2006-06-20)
Four genes have recently been proposed as candidates for dyslexia: dyslexia susceptibility 1 candidate 1 (DYX1C1), roundabout Drosophila homolog 1 (ROBO1), doublecortin domain-containing protein 2 (DCDC2) and KIAA0319. Each gene is implicated in global brain-development processes such as neural migration
Haiying Meng et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(47), 17053-17058 (2005-11-10)
DYX2 on 6p22 is the most replicated reading disability (RD) locus. By saturating a previously identified peak of association with single nucleotide polymorphism markers, we identified a large polymorphic deletion that encodes tandem repeats of putative brain-related transcription factor binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service