Skip to Content
Merck
All Photos(1)

Documents

D2691

Sigma-Aldrich

Monoclonal Anti-DC-SIGN1/DC-SIGN2 antibody produced in mouse

clone 120612, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-CD209/CD299, Anti-DC-SIGN Related, Anti-DC-SIGN/DC-SIGNR, Anti-Dendritic Cell-Specific ICAM-3 Grabbing Non-Integrin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

120612, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

flow cytometry: suitable
immunohistochemistry: 8-25 μg/mL using cell or in fixed tissue sections

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

DC-SIGN1 is a type II protein C-type lectin that binds mannose-rich molecules such as ICAM-3 and ICAM-2 in a calcium dependent manner. DC-SIGN1, a 44kDa molecule, is identical to the HIV-1 binding protein. Analysis of various tissues and cell lines suggests that DC-SIGN1 expression is restricted to dendritic cells, although there is evidence of expression in placenta, resting monocytes, and monocytic cell lines. DC-SIGN2 (DC-SIGNR), located on human chromosome 19p13.3, shares 73-80% amino acid identity with DC-SIGN1. DC-SIGN2 is polymorphic and is primarily transcribed in the liver and lymph nodes but not in monocyte-derived DC. Expression of DC-SIGN2 is restricted to endothelial cells derived from liver sinusoids, lymph nodes sinuses, and capillaries. Membrane and soluble forms of the protein have also been found to be expressed in placenta and some monocytic cell lines.

Specificity

Recognizes both human DC-SIGN1 and human DC-SIGN2 on transfected cells and reacts with human dendritic cells. The antibody does not react with parental mouse cells or irrelevant transfectants.

Immunogen

purified from NIH/3T3 cells transfected with recombinant human DC-SIGN2 (DC-SIGNR).

Application

Monoclonal Anti-DC-SIGN1/DC-SIGN2 antibody produced in mouse is suitable for flow cytometry and immunohistochemistry at a working concentration of 8-25ug/mL using cell or in fixed tissue sections.

Biochem/physiol Actions

DC-SIGN1 is involved in molecular interactions between dendritic cells and resting T-cells. DC-SIGN interacts with viruses such as X4, R5, X4R5 HIV-1 strains, HIV-2, and SIV. It captures HIV-1 through its interaction with the gp120 envelope glycoprotein and promotes efficient infection of CD4+ T cells that co-express chemokine receptors. DC-SIGN2 also binds mannose residues in a calcium dependent fashion, including ICAM-3 and HIV-1 gp120. It binds to human and simian immunodeficiency viruses and activates infection in trans.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Bashirova et al.
The Journal of experimental medicine, 193(6), 671-678 (2001-03-21)
The discovery of dendritic cell (DC)-specific intercellular adhesion molecule (ICAM)-3-grabbing nonintegrin (DC-SIGN) as a DC-specific ICAM-3 binding receptor that enhances HIV-1 infection of T cells in trans has indicated a potentially important role for adhesion molecules in AIDS pathogenesis. A
S Pöhlmann et al.
Journal of virology, 75(10), 4664-4672 (2001-04-20)
Dendritic cells (DCs) efficiently bind and transmit human immunodeficiency virus (HIV) to cocultured T cells and so may play an important role in HIV transmission. DC-SIGN, a novel C-type lectin that is expressed in DCs, has recently been shown to
S Pöhlmann et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(5), 2670-2675 (2001-02-28)
DC-SIGN, a C-type lectin expressed on the surface of dendritic cells (DCs), efficiently binds and transmits HIVs and simian immunodeficiency viruses to susceptible cells in trans. A DC-SIGN homologue, termed DC-SIGNR, has recently been described. Herein we show that DC-SIGNR
E J Soilleux et al.
Journal of immunology (Baltimore, Md. : 1950), 165(6), 2937-2942 (2000-09-07)
DC-SIGN is a C-type lectin, expressed on a dendritic cell subset. It is able to bind ICAM3 and HIV gp120 in a calcium-dependent manner. Here we report the genomic organization of DC-SIGN and map it to chromosome 19p13 adjacent to
B M Curtis et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(17), 8356-8360 (1992-09-01)
The binding of the human immunodeficiency virus (HIV) envelope glycoprotein gp120 to the cell surface receptor CD4 has been considered a primary determinant of viral tropism. A number of cell types, however, can be infected by the virus, or bind

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service