Skip to Content
Merck
All Photos(1)

Key Documents

D2293

Sigma-Aldrich

N-(2,4-Dinitrophenyl)-Pro-Leu-Gly-Leu-Trp-Ala-D-Arg amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H64N14O11
CAS Number:
Molecular Weight:
977.08
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

solubility

H2O: 2 mg/mL

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)CNC(=O)[C@H](CC(C)C)NC(=O)[C@@H]1CCCN1c2ccc(cc2N(=O)=O)N(=O)=O)C(=O)N[C@@H](Cc3c[nH]c4ccccc34)C(=O)N[C@@H](C)C(=O)N[C@H](CCCNC(N)=N)C(N)=O

Substrates

Fluorogenic substrate for matrix metalloproteinases (MMP-1 and MMP-9). The tryptophan fluorescence of the intact molecule is quenched by the dinitrophenyl moiety. Enzymatic cleavage of the substrate by collagenase or gelatinase results in an increased fluorescence.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G M McGeehan et al.
The Journal of biological chemistry, 269(52), 32814-32820 (1994-12-30)
The peptide substrate specificities of two matrix metalloproteinases (MMPs), interstitial collagenase (MMP-1), and 92-kDa gelatinase (MMP-9), have been examined. Starting with the parent substrate, Dnp-Pro-Leu-Gly approximately Leu-Trp-Ala-D-Arg-NH2, four separate substrate mixtures were synthesized at subsites P2(Leu) through P2'(Trp). These mixtures
S Upadhye et al.
Biochemical and biophysical research communications, 215(2), 474-482 (1995-10-13)
To correlate structural data on substrates of human fibroblast collagenase with their interaction with the enzyme, we have studied: Ac-PLG-s-LLG-O-ethyl ester (I), Dnp-PLGLWA(d-Arg)-NH2 (II), AcGPEGLRVG-O-ethyl ester (III) and Succ-GPLGP-O-amidomethylcoumaryl ester (IV). Peptides I and II represent collagenase cleavage sequences in
M S Stack et al.
The Journal of biological chemistry, 264(8), 4277-4281 (1989-03-15)
A fluorogenic substrate for vertebrate collagenase and gelatinase, Dnp-Pro-Leu-Gly-Leu-Trp-Ala-D-Arg-NH2, was designed using structure-activity data obtained from studies with synthetic inhibitors and other peptide substrates of collagenase. Tryptophan fluorescence was efficiently quenched by the NH2-terminal dinitrophenyl group, presumably through resonance energy
J Berman et al.
The Journal of biological chemistry, 267(3), 1434-1437 (1992-01-25)
A strategy is described for the rapid optimization of kcat/Km for protease substrates. Selected positions of a given peptide substrate sequence are varied through synthesis with mixtures of amino acids. Incubation of the resulting peptide mixture with the enzyme of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service