All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H22N2O4S · C6H13N
CAS Number:
Molecular Weight:
449.61
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
storage temp.
−20°C
SMILES string
NC1CCCCC1.CC(C)C(NS(=O)(=O)c2cccc3c(cccc23)N(C)C)C(O)=O
InChI
1S/C17H22N2O4S.C6H13N/c1-11(2)16(17(20)21)18-24(22,23)15-10-6-7-12-13(15)8-5-9-14(12)19(3)4;7-6-4-2-1-3-5-6/h5-11,16,18H,1-4H3,(H,20,21);6H,1-5,7H2
InChI key
XAAKQPYBZYQKNA-UHFFFAOYSA-N
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Deying Kong et al.
Electrophoresis, 39(2), 363-369 (2017-09-12)
Pillar[n]arenes have achieved much interest in material chemistry and supramolecular chemistry due to unusual pillar shape structure and high selectivity toward guest. However, pillar[n]arenes have not yet been applied in capillary electrochromatography. This work at first time reports that carboxylatopillar[5]arene
Chromatograms
application for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service