Skip to Content
Merck
All Photos(1)

Key Documents

D0939

Sigma-Aldrich

2′,5′-Dideoxyadenosine 3′-triphosphate tetrasodium salt

>91% (HPLC)

Synonym(s):

2′,5′-Dideoxy-3′-ATP, 2′,5′-Dideoxyadenosine 3′-triphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N5O11P3Na4
CAS Number:
Molecular Weight:
563.11
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

>91% (HPLC)

form

solid

color

off-white, powder

solubility

H2O: 24 mg/mL

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].[Na+].C[C@H]1O[C@H](C[C@@H]1OP([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O)n2cnc3c(N)ncnc23

InChI

1S/C10H16N5O11P3.4Na/c1-5-6(24-28(19,20)26-29(21,22)25-27(16,17)18)2-7(23-5)15-4-14-8-9(11)12-3-13-10(8)15;;;;/h3-7H,2H2,1H3,(H,19,20)(H,21,22)(H2,11,12,13)(H2,16,17,18);;;;/q;4*+1/p-4/t5-,6+,7-;;;;/m1..../s1

InChI key

BKMXLPGGJURCPM-DNGRLYOHSA-J

Biochem/physiol Actions

Potent inhibitor of adenylyl cyclase. Not cell-permeable. IC50 = 40 nM in detergent-dispersed rat brain preparation.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Adenylyl cyclases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Désaubry et al.
The Journal of biological chemistry, 271(24), 14028-14034 (1996-06-14)
The synthesis of a number of adenine nucleoside 3'-polyphosphates has been devised via a phosphotriester approach that combines the method of alkoxide activation with the use of 2,2,2-tribromoethyl phosphoromorpholinochloridate as a phosphorylating agent. The family of compounds included 3'ADP, 3'ATP
L Désaubry et al.
The Journal of biological chemistry, 271(5), 2380-2382 (1996-02-02)
2',5'-Dideoxyadenosine 3'-di- and triphosphates were tested as inhibitors of brain adenylyl cyclases. With an IC50 approximately 40 nM, 2',5'-dideoxy-3'-ATP is the most potent nonprotein synthetic regulator of adenylyl cyclases thus far described. Neither 2',5'-dideoxy-3'-ADP nor 2',5'-dideoxy-3'-ATP inhibited activity by competition
Adenylyl Cyclases.
Johnson, R.A. et al.
Encyclopedic Reference of Molecular Pharmacology (2003)
L Désaubry et al.
The Journal of biological chemistry, 273(38), 24972-24977 (1998-09-12)
2'-Deoxyadenosine 3'-tetraphosphate (2'-deoxy-3'-A4P) and 2', 5'-dideoxyadenosine 3'-tetraphosphate (2',5'-dideoxy-3'-A4P) were synthesized, and their effects were tested on crude and purified forms of native adenylyl cyclases isolated from brain. Syntheses combined the method of alkoxide activation with the use of tribromoethyl phosphoromorpholino-chloridate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service