CWDS-IF790
ColorWheel® Antibody-Ready iFluor 790 Dye
for use with ColorWheel® Antibody (Required, sold separately
)
Synonym(s):
iFluor 790
About This Item
Recommended Products
product line
ColorWheel®
Quality Level
form
lyophilized
purified by
ion-exchange chromatography
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
flow cytometry: suitable
fluorescence
λex 787 nm; λem 812 nm
compatibility
for use with ColorWheel® Antibody (
Required, sold separately
)greener alternative category
, Aligned
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
unmodified
General description
iFluor 790 is designed to label proteins and other biomolecules with near infrared fluorescence. iFluor 790 conjugates exhibit excitation (787 nm) and emission (812 nm) spectra similar to that of indocyanine green (ICG) and the IRDye® 800 dye and are well separated from commonly used far-red fluorophores such as Cy5, Cy7, and allophycocyanin (APC). Hence, it is suitable for multicolor analysis. It can also be used in small animals for in-vivo imaging applications.
Specificity
Application
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
CWD-IF790-25UL - ColorWheel® Antibody-Ready iFluor 790 Dye Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.
CWD-FINISH-25UL - ColorWheel® Finishing Buffer Lyophilized from PBS. Contains no biocide or preservatives, such as azide.
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service