Skip to Content
Merck
All Photos(7)

Key Documents

CWA-1036

Sigma-Aldrich

Anti-Human CD20 (2H7) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, sold separately)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2H7, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 33.08 kDa

species reactivity

human

species reactivity (predicted by homology)

nonhuman primates

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, sold separately)

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone 2H7 is a mouse monoclonal antibody that detects CD20. It targets an epitope within 16 amino acids from the second extracellular domain.

Immunogen

Human tonsillar B cells.

Application

Quality Control Testing:

Evaluated by Flow Cytometry in Raji cells with Fc block.

Staining of one million Human Raji cells with Fc block was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1036, Anti-Human CD20 (2H7) ColorWheel® Dye-Ready mAb and Cat. No. CWD-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or the equivalent amount of Cat. No. CWA-1010 Mouse IgG2b isotype control and Cat. No. CWD-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

B-lymphocyte antigen CD20 (UniProt: P11836; also known as B-lymphocyte surface antigen B1, Bp35, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, CD20) is encoded by the MS4A1 (also known as CD20) gene (Gene ID: 931) in human. CD20 is a multi-pass homotetrameric, non-glycosylated membrane protein that is expressed on the surface of normal and malignant B-lymphocytes. CD20 expression is reportedly initiated at the pre-B cell stage of development and remains present until terminal differentiation into a plasma cell at which time it is lost. It has three cytoplasmic domains, four transmembrane domains, and two extracellular domains. Its both N- and C-termini reside within the cytoplasm. CD20 plays a role in the regulation of cellular calcium influx necessary for the development, differentiation, and activation of B-lymphocytes. It functions as a store-operated calcium channel component. It undergoes phosphorylation by PKC on serine and threonine residues in resting cells and these phosphorylations are reported to be higher in proliferating malignant B cells than in resting cells. CD20 is considered as an important therapeutic target in the treatment of B cell malignancies and various antibodies have been developed that bind to different epitopes . (Ref.: Pavlasova, G., and Mraz, M. (2020). Haematologica. 105(6); 1494-1506; Klein, C., et al. (2013). MAbs. 5(1); 22 33).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL of PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service