Key Documents
CLS766116
Pyrex® pennyhead stopper
stopper joint: 16
Synonym(s):
glass stopper, glass stoppers, stoppers, Corning® 7660
About This Item
UNSPSC Code:
41122500
NACRES:
NB.43
Pricing and availability is not currently available.
material
glass
Joint
joint: stopper16
manufacturer/tradename
Corning 7661-16
General description
Ground zone length indicates its greatest diameter (mm).
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)