Skip to Content
Merck
All Photos(1)

Documents

CLS7058150

Pyrex® VISTA volumetric flask, Class B

capacity 50 mL

Synonym(s):

flasks glass, volumetric flasks, volumetric glass

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

material

borosilicate glass
long

sterility

non-sterile

feature

class B
closure type Snap cap

packaging

case of 12
pack of 6

manufacturer/tradename

Corning 70581-50

capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

General description

These Pyrex VISTA Class B volumetric flasks have capacity tolerances that are twice those specified for Class A volumetric ware. The graduation line is sharp and permanent; large blue clock letters make inscriptions easy to read. Snap-caps are supplied with all sizes.

Legal Information

PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service