Skip to Content
Merck
All Photos(1)

Key Documents

CLS705811L

Pyrex® VISTA volumetric flask, Class B

capacity 1 L

Synonym(s):

flasks glass, volumetric flasks, volumetric glass

Sign Into View Organizational & Contract Pricing

Select a Size

6 EA
R$1,646.00

R$1,646.00


Check Cart for Availability


Select a Size

Change View
6 EA
R$1,646.00

About This Item

UNSPSC Code:
41121804

R$1,646.00


Check Cart for Availability

material

borosilicate glass
long

sterility

non-sterile

feature

class B
closure type Snap cap

packaging

case of 6

manufacturer/tradename

Corning 70581-1L

capacity

1 L

Looking for similar products? Visit Product Comparison Guide

General description

These Pyrex VISTA Class B volumetric flasks have capacity tolerances that are twice those specified for Class A volumetric ware. The graduation line is sharp and permanent; large blue clock letters make inscriptions easy to read. Snap-caps are supplied with all sizes.

Legal Information

PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service