Key Documents
CLS64062LFO
PYREX® replacement Squibb separatory funnel w/o valve and stopper
pack of 1 ea
Synonym(s):
equipment replacement accessory
About This Item
UNSPSC Code:
41121809
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Corning 6406-2LFO
capacity
2 L
volume
2 L
20000 mL
Related Categories
General description
These economy Squibb separatory funnel replacements do not have a Polytetrafluorethylene (PTFE) valve assembly or polyethylene stopper.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)