CLS4624
Corning® microcarriers
untreated, weight 100 g
Synonym(s):
lab equipment accessory
Select a Size
R$4,352.00
Select a Size
About This Item
R$4,352.00
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Corning 4624
bead size
125-212 μm
weight
100 g
density
~1.026 g/mL
Looking for similar products? Visit Product Comparison Guide
General description
Corning microcarriers with Corning CellBIND are available in 100 g and 500 g bottles that can be fitted with Corning aseptic transfer caps to prevent adventitious contamination during bioreactor loading. The transfer caps are available with either a male Luer lock quick connector or an MPC quick connect. Microcarriers and aseptic transfer caps are sold separately.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service