CLS432047
Corning® CoolRack®
M15-PF, capacity 15 x 1.5 mL microfuge tubes, tapered wells for conical tubes, gray
Synonym(s):
Corning Coolcell accessory
Select a Size
R$2,106.00
Select a Size
About This Item
R$2,106.00
Recommended Products
material
foamed PE
feature
wells 15
Holds 15 x 1.5 mL tubes
packaging
pkg of 1
manufacturer/tradename
Corning 432047
technique(s)
cryopreservation: suitable
L × W × H
10.2 cm × 6.4 cm × 3.8 cm
size
17.8 mm , column spacing
17.8 mm , row spacing
well depth
35.3 mm
well diam.
11.1 mm
color
gray
Looking for similar products? Visit Product Comparison Guide
General description
- Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
- All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
- Certain CoolRack modules are SBS-compatible.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service