Skip to Content
Merck
All Photos(1)

Key Documents

CLS432027

Corning® CoolBox, 2XT System

workstation, double capacity, orange

Synonym(s):

Staining, Staining Accessories

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$7,961.00

R$7,961.00


Check Cart for Availability


Select a Size

Change View
1 EA
R$7,961.00

About This Item

UNSPSC Code:
41123300
NACRES:
NB.12

R$7,961.00


Check Cart for Availability

feature

workstation, double capacity

packaging

pkg of 1

manufacturer/tradename

Corning 432027

size

21.0 cm × 14.3 cm × 13.4 cm, Internal Dimensions
26.5 cm × 20.0 cm × 15.0 cm, External Dimensions

color

orange

Looking for similar products? Visit Product Comparison Guide

General description

Corning CoolBox 2XT systems are bench top cooling workstations that accomodate two CoolRacks and provide sample cooling or freezing without ice, electricity or batteries. CoolBox systems are versatile and accommodate a variety of sample formats and temperatures. The modular design enables the use of CoolRack® and CoolSink® thermo-conductive sample modules to hold microfuge tubes, cryogenic vials, PCR tubes or plates, assay plates and more.

  • Includes two (2) XT Cooling Cores for 0.5° to 4°C cooling
  • Accommodates two (2) sample modules
  • Includes a magnetized collar and a 1° to 8°C temperature indicator for visual assurance of proper temperature
  • To use with any 30 place Corning CoolRack Tubes Module
  • The internal cooling or freezing cores provide 0.5° to 4°C for over 10 hours with lid open and over 16 hours with lid closed
  • CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.

Legal Information

CoolBox is a trademark of Corning, Inc.
CoolRack is a registered trademark of Corning, Inc.
CoolSink is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service