Skip to Content
Merck
All Photos(1)

Key Documents

CLS4219

Corning® Step-R Repeating Pipettor Adapter for 50 mL Syringe Tips

non-sterile

Synonym(s):

repeater

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$40.00

R$40.00


Check Cart for Availability


Select a Size

Change View
1 EA
R$40.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

R$40.00


Check Cart for Availability

material

red polypropylene

sterility

non-sterile

feature

Autoclavable at 121°C, 25 minutes

packaging

pack of 1 ea

manufacturer/tradename

Corning 4219

Related Categories

General description

Suitable for use with 50 mL syringe tips.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service